Diamond DNA Ladder 100 to 10 Kbp, Rockland Immunochemicals
Catalog # RLMB-205-0500
Supplier: Rockland Immunochemical
CAS Number:
Specifications
- Description:Diamond DNA Ladder 100-10Kbp
- Size:500 µL
- Shipping Temperature:Wet Ice
- Cat. No.:RLMB-205-0500
- Supplier no.:MB-205-0500
Specifications
About this item
Diamond DNA Ladder 100-10Kbp
- comprised of PCR products and double-stranded DNA
- digested with restriction enzymes, phenol extracted, equilibrated to 10 mM Tris-HCl (pH 8.0) and 10mM EDTA
Diamond Ready-to-Use DNA Ladder 100-10Kbp is a unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 19 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 100-10,000 base pairs. The 500, 1.5K and 3K bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.86 μg a load) for approximating the mass of DNA in comparably intense samples of similar size.
Diamond Ready-to-Use DNA Ladder 100-10Kbp has a suggested load volume is 5 µL/well.