Order Entry
United States
ContactUsLinkComponent
Clostridium histolyticum Collagenase Type 4
Clostridium histolyticum Collagenase Type 4
Catalog # RLMB-125-0050
CAS Number:  
Clostridium histolyticum Collagenase Type 4
Catalog # RLMB-125-0050
Supplier Number:  MB-125-0050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
    Clostridium histolyticum
  • Size:
    50 mg
  • Environmentally Preferable:
  • Shipping Conditions:
    Ambient
  • Enzyme Name:
    Collagenase Type 4
  • Enzyme Synonyms:
    colH, colG
  • Concentration:
    ≥160 CDU/mg (dry weight)
  • Cat. No.:
    RLMB-125-0050
  • Supplier no.:
    MB-125-0050

Specifications

About this item

Collagenase Type 4

  • Crude collagenases are widely used in enzymatic primary cell isolation and tissue dissociation procedures.
  • Most researchers employ either crude collagenase preparations such as Types 1, 2, 3, and 4 or chromatographically purified collagenase; the latter usually combined with secondary enzymes such as elastase, hyaluronidase, etc.
  • For best results, the precise mixture of proteolytic activities must be tailored to the tissue to be dissociated.
  • Collagenase is ideal for researches focused in Stem Cell and Biomarker Research.

Crude collagenase preparations contain several isoforms of two different collagenases, a sulfhydryl protease, clostripain, a trypsin-like enzyme, and an aminopeptidase. This combination of collagenolytic and proteolytic activities is effective at breaking down intercellular matrices, the essential part of tissue dissociation. One component of the complex is a hydrolytic enzyme which degrades the helical regions in native collagen preferentially at the Y-Gly bond in the sequence Pro-Y-Gly-Pro, where Y is most frequently a neutral amino acid. This cleavage yields products susceptible to further peptidase digestion. Crude collagenase is inhibited by metal chelating agents such as cysteine, EDTA or o-phenanthroline but not DFP. It is also inhibited by α2-macroglobulin, a large plasma glycoprotein. Ca2+ is required for enzyme activity. Particular enzymatic profiles of each collagenase have been correlated with the tissues from which the cells for study were obtained (or with the uses to which the cells are put) and as a result of the correlations several types of crude collagenases have been established.

Collagenase Type 4 contain lower tryptic activity levels to limit damage to membrane proteins and receptors but with normal to above normal collagenase activity. It is suggested for pancreatic islet primary isolation. Collagenase is typically used at concentrations from 0.05 % to 0.5 % (w/v) in balanced salt solutions such as Hank’s, Earle’s and others. For best results the precise mixture of collagenase and proteolytic activities must be tailored to the tissue to be dissociated. Specific conditions for reactivity should be optimized by the end user.