Micrococcal Nuclease S7
Catalog # 75934-884
Supplier: Rockland Immunochemical
CAS Number:
Specifications
- Size:15 KU
- Shipping Conditions:Wet ice
- Enzyme Name:Micrococcal Nuclease S7
- Cat. No.:75934-884
- Supplier no.:MB-116-0015
Specifications
About this item
Micrococcal Nuclease S7
- ≥6,000 units per mg protein.
- Chromatographically purified to be essentially homogeneous chromatographically and electrophoretically (SDS-PAGE).
Micrococcal nuclease catalyzes cleavage of both DNA and RNA to yield 3'-nucleotides. It exhibits exo- and endo-5'-phosphodiesterase activities. The enzyme catalyzes preferential endohydrolysis of the RNA and DNA at sites rich in adenylate or uridylate and deoxyadenylate or thymidylate. The enzyme has a molecular weight of 16,807 daltons and is calcium dependent. The pH optimum is 9.2 but varies depending upon the concentration of ionized calcium present.