Order Entry
United States
ContactUsLinkComponent
Rat CCL7 - MCP-3 ELISA Kit, Rockland Immunochemicals
Rat CCL7 - MCP-3 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-488
CAS Number:  
Rat CCL7 - MCP-3 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-488
Supplier Number:  KOA0774
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Description:
    Rat CCL7 - MCP-3 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-488

Specifications

About this item

Rat CCL7 - MCP-3 AccuSignal ELISA Kit

  • Natural and recombinant rat CCL7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Q24-P97

Chemokine(C-C motif) ligand 7(CCL7) is a small cytokine known as a chemokine that was previously called monocyte-specific chemokine 3(MCP3). It belongs to the C-C chemokine family. By fluorescence in situ hybridization, mapped the MCP3 gene to chromosome 17q11.2-q12. MCP3 was identified as a physiologic substrate of gelatinase A. Cleaved MCP3 binds to CC-chemokine receptors-1, -2, and -3, but no longer induces calcium fluxes or promotes chemotaxis, and instead acts as a general chemokine antagonist that dampens inflammation, suggested that matrix metalloproteinases are both effectors and regulators of the inflammatory response.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat CCL7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat CCL7 standard solution and each sample be measured in duplicate.