Order Entry
United States
ContactUsLinkComponent
Human Galectin-3BP ELISA Kit, Rockland Immunochemicals
Human Galectin-3BP ELISA Kit, Rockland Immunochemicals
Catalog # 76235-938
CAS Number:  
Human Galectin-3BP ELISA Kit, Rockland Immunochemicals
Catalog # 76235-938
Supplier Number:  KOA0729
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human Galectin-3BP ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-938

Specifications

About this item

Human Galectin-3BP AccuSignal ELISA Kit

  • Natural and recombinant human Galectin-3BP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: V19-D585

Galectin-3-binding protein is a protein that in humans is encoded by the LGALS3BP gene. The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell–cell and cell–matrix interactions. Using fluorescence in–situ hybridization, the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds to galectin 1.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Galectin-3BP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human Galectin-3BP standard solution and each sample be measured in duplicate.