- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse CCL9 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-916
Mouse CCL9 AccuSignal ELISA Kit
- Natural and recombinant mouse CCL9 .
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: Q22-Q122
Chemokine(C-C motif) ligand 9(CCL9), also called MIP-1 gamma and MRP-2, is a small cytokine belonging to the CC chemokine family. CCL9 which found around Peyer's patches is secreted by follicle-associated epithelium(FAE), and it can attract dendritic cells that possess the cell surface molecule CD11b and the chemokine receptor CCR1. CCL9 is constitutively expressed in macrophages and myeloid cells. CCL9 can activate osteoclasts through its receptor CCR1(the most abundant chemokine receptor found on osteoclasts) suggesting an important role for CCL9 in bone resorption.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml mouse CCL9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse CCL9 standard solution and each sample is measured in duplicate.