Specifications
- Primary antibody reactivity:Human
- Description:Human soluble CD6 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-908
Specifications
About this item
Human soluble CD6 AccuSignal ELISA Kit
- Natural and recombinant human CD6.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: H18-E398
CD6(Cluster of Differentiation 6) is a human protein encoded by the CD6 gene. It is mapped to 11q12.2. CD6 is expressed predominantly in peripheral T cells and mature medullary thymocytes. This gene encodes a protein found on the outer membrane of T-lymphocytes as well as some other immune cells. The encoded protein contains three scavenger receptor cysteine-rich(SRCR) domains and a binding site for an activated leukocyte cell adhesion molecule. The gene product is important for continuation of T cell activation. What's more, this gene may be associated with susceptibility to multiple sclerosis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human CD6 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD6 standard solution and each sample be measured in duplicate.