Order Entry
United States
ContactUsLinkComponent
Human CCL15 ELISA Kit, Rockland Immunochemicals
Human CCL15 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-802
CAS Number:  
Human CCL15 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-802
Supplier Number:  KOA0640
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CCL15 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-802

Specifications

About this item

Human CCL15 AccuSignal ELISA Kit

  • Natural and recombinant human CCL15.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Q22-I113

Chemokine(C-C motif) ligand 15(CCL15) is a small cytokine belonging to the CC chemokine family that is also known as leukotactin-1, MIP5 and HCC-2. CCL15 is expressed in liver, small intestine, colon, and in certain leukocytes and macrophages of the lung. It is chemotactic for neutrophils, monocytes, and lymphocytes and elicits its effects by binding to cell surface chemokine receptors like CCR1 and CCR3. The human CCL15 gene spans four exons and is located in a head-to-tail orientation on chromosome 17 with the gene of another CC chemokine known as CCL14.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human CCL15 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CCL15 standard solution and each sample be measured in duplicate.