Specifications
- Primary antibody reactivity:Human
- Description:Human Kallikrein 14 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-700
Specifications
About this item
Human Kallikrein 14 AccuSignal ELISA Kit
- Natural and recombinant human Kallikrein 14.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q19-M248
Kallikrein-14 is a protein that in humans is encoded by the KLK14 gene. Kallikrein-14 belongs to the kallikrein subgroup of serine proteases, which have diverse physiologic functions in many tissues. Kallikrein-14 gene is identified in the kallikrein gene cluster on chromosome 19q13.4. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. Kallikrein-14 may activate or inactivate the proteinase-activated receptors F2R, F2RL1 and F2RL3 and other kallikreins including KLK1, KLK3, KLK5 and KLK11.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human Kallikrein 14 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates, serum, plasma(heparin, EDTA) or tissue homogenates to each empty well. It is recommended that each human Kallikrein 14 standard solution and each sample be measured in duplicate.