Order Entry
United States
ContactUsLinkComponent
Rat sE-Selectin ELISA Kit, Rockland Immunochemicals
Rat sE-Selectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-430
CAS Number:  
Rat sE-Selectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-430
Supplier Number:  KOA0571
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Rat
  • Target protein:
    sE-selectin
  • Description:
    Rat sE-Selectin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-430

Specifications

About this item

Rat sE-Selectin AccuSignal ELISA Kit

  • Natural and recombinant rat sE-Selectin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: W22-P494

E-selectin, also called endothelial-leukocyte adhesion molecule-1(ELAM-1), is a cell surface glycoprotein expressed by cytokine-activated endothelium that mediates the adhesion of blood neutrophils.1 An increased expression of E-selectin has been observed in the arterial endothelium interacting with lymphocytes and macrophages in human atherosclerotic lesions.2 E-selectin plays a critical role in mediating tissue-specific homing of T cells into skin, and of primitive hematopoietic progenitor cells(HPCs) into bone marrow(BM).3 A structurally and functionally related group of genes, lymph node homing receptor(LHR), granule membrane protein 140(GMP-140), and endothelial leukocyte adhesion molecule 1(ELAM-1) are shown to constitute a gene cluster on mouse and human chromosome 1.4

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml rat E-Selectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, citrate) to each empty well. It is recommended that each rat E-Selectin standard solution and each sample be measured in duplicate.