Order Entry
United States
ContactUsLinkComponent
Mouse GDNF ELISA Kit, Rockland Immunochemicals
Mouse GDNF ELISA Kit, Rockland Immunochemicals
Catalog # 76237-428
CAS Number:  
Mouse GDNF ELISA Kit, Rockland Immunochemicals
Catalog # 76237-428
Supplier Number:  KOA0570
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse GDNF ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-428

Specifications

About this item

Mouse GDNF AccuSignal ELISA Kit

  • Natural and recombinant mouse GDNF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: S78-I211

Glial cell line-derived neurotrophic factor(GDNF) is a glycosylated, disulfide-bonded homodimer that is a distantly related member of the transforming growth factor-beta superfamily.1 GDNF, is a potent neurotrophic factor that promotes the survival of dopaminergic neurones in cultures including embryonic neuronal cultures.2 GDNF, in addition to its potential role in the differentiation and survival of central nervous system neurons, has profound effects on kidney organogenesis and the development of the peripheral nervous system.3 GDNF may have utility in the treatment of Parkinson's disease, which is marked by progressive degeneration of midbrain dopaminergic neurons.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse GDNF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse GDNF standard solution and each sample be measured in duplicate.