Order Entry
United States
ContactUsLinkComponent
Human Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Human Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-416
CAS Number:  
Human Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-416
Supplier Number:  KOA0564
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human Tissue factor - F3 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-416

Specifications

About this item

Human Tissue factor - F3 AccuSignal ELISA Kit

  • Natural and recombinant human Tissue factor.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S33-E251

Tissue factor, also called platelet tissue factor, factor III, thrombokinase, or CD142 is a protein present in sub endothelial tissue and leukocytes necessary for the initiation of thrombin formation from the zymogene prothrombin. An incorrect synonym is thromboplastin. the F3 gene was mapped to 1pter-p21 by study of somatic cell hybrids with a species-specific sensitive chromogenic assay. This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human Tissue factor standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA, citrate) cell lysates or urine to each empty well. It is recommended that each human Tissue factor standard solution and each sample be measured in duplicate.