Order Entry
United States
ContactUsLinkComponent
Mouse RBP4 ELISA Kit, Rockland Immunochemicals
Mouse RBP4 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-358
CAS Number:  
Mouse RBP4 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-358
Supplier Number:  KOA0499
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse RBP4 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-358

Specifications

About this item

Mouse RBP4 AccuSignal ELISA Kit

  • Natural and recombinant mouse RBP4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E19-L201

Retinol binding protein 4, plasma, also known as RBP4, belongs to the lipocalin family and is the specific carrier for retinol(vitamin A alcohol) in the blood. It is protein that it is encoded by the RBP4 gene. RBP4 gene resides just centromeric of the cluster of CYP2C genes on 10q24. The mouse Rbp4 locus is closely linked and just proximal to the locus for phenobarbital-inducible cytochrome P450-2c(Cyp-2c) at the distal end of chromosome 19. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin, which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells. The standard product used in this kit are recombinant mouse B7-1/CD80, D37—K245, consisting of dimer acids with two single straned. The standard used in this kit is recombinant protein, with E19-L201 aa sequence, the molecular weight is 22kda.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml mouse RBP4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin , EDTA) or urine to each empty well. We recommend that each mouse RBP4 standard solution and each sample is measured in duplicate.