Order Entry
United States
ContactUsLinkComponent
Mouse MER - MERTK ELISA Kit, Rockland Immunochemicals
Mouse MER - MERTK ELISA Kit, Rockland Immunochemicals
Catalog # 76236-908
CAS Number:  
Mouse MER - MERTK ELISA Kit, Rockland Immunochemicals
Catalog # 76236-908
Supplier Number:  KOA0482
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse MER - MERTK ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-908

Specifications

About this item

Mouse MER - MERTK AccuSignal ELISA Kit

  • Natural and recombinant mouse MER.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: E23-F498

Proto-oncogene tyrosine-protein kinase MER, also called MERTK is an enzyme that in humans is encoded by the MERTK gene. This gene is a member of the MER/AXL/TYRO3 receptor kinase family and encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type(immunoglobulin-like) domains, and one tyrosine kinase domain. This gene is mapped to 2q13. MERTK signaling plays a role in various processes such as macrophage clearance of apoptotic cells, platelet aggregation, cytoskeleton reorganization and engulfment. Functions in the retinal pigment epithelium(RPE) as a regulator of rod outer segments fragments phagocytosis. This gene also plays an important role in inhibition of Toll-like receptors(TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signaling SOCS1 and SOCS3.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml mouse MER standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, cell lysates, tissue homogenates or serum to each empty well. We recommend that each mouse MER standard solution and each sample is measured in duplicate.