Specifications
- Primary antibody reactivity:Human
- Description:Human C-MET - HGFR ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-298
Specifications
About this item
Human C-MET - HGFR AccuSignal ELISA Kit
- Natural and recombinant human C-MET.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E25-R307(alpha)&S308-T932(beta)
C-Met(MET or MNNG HOS Transforming gene) is a proto-oncogene that encodes a protein known as hepatocyte growth factor receptor(HGFR). MET proto-oncogene has a total length of 125,982 bp, and it is located in the 7q31 locus of chromosome 7. MET is a membrane receptor that is essential for embryonic development and wound healing. Activation of MET triggers mitogenesis, and morphogenesis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human C-MET standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human C-MET standard solution and each sample be measured in duplicate.