ContactUsLinkComponent
Human IL7R - CD127 ELISA Kit, Rockland Immunochemicals
  76236-896
 :  
Human IL7R - CD127 ELISA Kit, Rockland Immunochemicals
  76236-896
 :  KOA0415
 :  
  • Primary antibody reactivity:
    Human
  • Description:
    Human IL7R - CD127 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-896
Human IL7R - CD127 AccuSignal ELISA Kit
  • Natural and recombinant human IL7R.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E21-I262
The interleukin-7 receptor(IL7R) is a protein found on the surface of cells. It is mapped to 5p13.2. The protein encoded by this gene is a receptor for interleukine 7(IL7). The function of this receptor requires the interleukin 2 receptor, gamma chain(IL2RG), which is a common gamma chain shared by the receptors of various cytokines, including interleukine 2, 4, 7, 9, and 15. IL7R has been shown to play a critical role in the development of immune cells called lymphocytes-specifically in a process known as V(D)J recombination. This protein is also found to control the accessibility of a region of the genome that contains the T-cell receptor gamma gene, by STAT5 and histone acetylation. What's more, IL7R antagonism is efficacious in treatment of EAE through its effects on Th17 cells and is a potential treatment for MS.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human IL7R standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human IL7R standard solution and each sample be measured in duplicate.