Order Entry
United States
ContactUsLinkComponent
Human CD40 - TNFRSF5 ELISA Kit, Rockland Immunochemicals
Human CD40 - TNFRSF5 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-272
CAS Number:  
Human CD40 - TNFRSF5 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-272
Supplier Number:  KOA0407
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    CD40
  • Description:
    Human CD40 - TNFRSF5 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-272

Specifications

About this item

Human CD40 - TNFRSF5 AccuSignal ELISA Kit

  • Natural and recombinant human CD40.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E21-R193

CD40 is a cell surface receptor that is expressed on the surface of all mature B cells, most mature B-cell malignancies, and some early B-cell acute lymphocytic leukemias, but is not expressed on plasma cells. The CD40 gene is localized to the long arm of chromosome 20, bands q12-q13.2. The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human CD40 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA), saliva, urine or human milk to each empty well. It is recommended that each human CD40 standard solution and each sample be measured in duplicate.