Order Entry
United States
ContactUsLinkComponent
Human CA9 ELISA Kit, Rockland Immunochemicals
Human CA9 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-250
CAS Number:  
Human CA9 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-250
Supplier Number:  KOA0392
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CA9 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-250

Specifications

About this item

Human CA9 AccuSignal ELISA Kit

  • Natural and recombinant human CA9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: P59-D414

Carbonic anhydrase 9(CA9/CAIX) is an enzyme that in humans is encoded by the CA9 gene. Carbonic anhydrases(CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. This gene is mapped to 9p13.3. CA IX is a transmembrane protein and the only tumor-associated carbonic anhydrase isoenzyme known. It is expressed in all clear-cell renal cell carcinoma, but is not detected in normal kidney or most other normal tissues. It may be involved in cell proliferation and transformation.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human CA9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA, citrate) or urine to each empty well. It is recommended that each human CA9 standard solution and each sample be measured in duplicate.