Order Entry
United States
ContactUsLinkComponent
Mouse Prolactin ELISA Kit, Rockland Immunochemicals
Mouse Prolactin ELISA Kit, Rockland Immunochemicals
Catalog # 76236-874
CAS Number:  
Mouse Prolactin ELISA Kit, Rockland Immunochemicals
Catalog # 76236-874
Supplier Number:  KOA0364
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Prolactin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-874

Specifications

About this item

Mouse Prolactin AccuSignal ELISA Kit

  • Natural and recombinant mouse Prolactin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: L32-C228

Prolactin, also calle PRL, is a protein that in humans is encodes by the PRL gene. This gene is mapped to 6p22.3. This gene encodes the anterior pituitary hormone prolactin. It may play a role in cell survival by suppressing apoptosis, and it is essential for lactation. Prolactin also acts in a cytokine-like manner and as an important regulator of the immune system. Prolactin has important cell cycle related functions as a growth-, differentiating- and anti-apoptotic factor. As a growth factor binding to cytokine like receptors it has also profound influence on hematopoiesis, angiogenesis and is involved in the regulation of blood clotting through several pathways.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse Prolactin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, citrate) to each empty well. It is recommended that each mouse Prolactin standard solution and each sample be measured in duplicate.