Order Entry
United States
ContactUsLinkComponent
 
Mouse Resistin ELISA Kit, Rockland Immunochemicals
Catalog # 76236-858
CAS Number:  
Mouse Resistin ELISA Kit, Rockland Immunochemicals
Catalog # 76236-858
Supplier Number:  KOA0356
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Resistin
  • Description:
    Mouse Resistin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-858

Specifications

About this item

Mouse Resistin AccuSignal ELISA Kit

  • Natural and recombinant mouse Resistin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S21-S114

Resistin is an adipose-derived hormone postulated to link adiposity to insulin resistance. Type 2 diabetes, characterized by target-tissue resistance to insulin, is epidemic in industrialized societies and is strongly associated with obesity. Adipocytes secrete a unique signalling molecule, which called resistin(for resistance to insulin), that may be the hormone potentially links obesity to diabetes.Resistin is overexpressed in mouse adipose tissue of obese individuals and is likely to modulate insulin sensitivity. Resistin is, therefore, a candidate gene for insulin resistance.The standard product used in this kit is mouse Resistin with the molecular mass of 51-53KDa. The detected Resistin includes zymogen and active enzyme.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse Resistin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin) to each empty well. We recommend that each mouse Resistin standard solution and each sample is measured in duplicate.