Order Entry
United States
ContactUsLinkComponent
Rat Rantes ELISA Kit, Rockland Immunochemicals
Rat Rantes ELISA Kit, Rockland Immunochemicals
  76237-170
 :  
Rat Rantes ELISA Kit, Rockland Immunochemicals
  76237-170
 :  KOA0295
 :  

 

  • Assay Type:
    Sandwich
  • Description:
    Rat Rantes ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-170

 

 

Rat Rantes AccuSignal ELISA Kit

  • Natural and recombinant rat Rantes.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S24-S91

Rantes(regulated on activation normal T cell expressed and secreted) is one of the natural ligands for the chemokine receptor CCR5 and potently suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Peripheral blood mononuclear cells or CD4(+) lymphocytes obtained from different individuals have wide variations in their ability to secrete RANTES. The Rantes gene product is predicted to be 10 kDa and, after cleavage of the signal peptide, approximately 8 kDa. Of the 68 residues, 4 are cysteines, and there are no sites for N-linked glycosylation. Rantes is expressed by cultured T cell lines that are Ag specific and growth factor dependent.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat Rantes standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each rat Rantes standard solution and each sample be measured in duplicate.