Order Entry
United States
ContactUsLinkComponent
Human OSM - Oncostatin M ELISA Kit, Rockland Immunochemicals
Human OSM - Oncostatin M ELISA Kit, Rockland Immunochemicals
Catalog # 76237-594
CAS Number:  
Human OSM - Oncostatin M ELISA Kit, Rockland Immunochemicals
Catalog # 76237-594
Supplier Number:  KOA0283
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    OSM
  • Description:
    Human OSM - Oncostatin M ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-594

Specifications

About this item

Human OSM - Oncostatin M AccuSignal ELISA Kit

  • Natural and recombinant human OSM.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A26R-220

OSM(Oncostatin M) is a protein that in humans is encoded by the OSM gene. OSM is a pleiotropic cytokine that belongs to the interleukin 6 group of cytokines. The OSM gene is mapped on 22q12.2. Of these cytokines it most closely resembles leukemia inhibitory factor(LIF) in both structure and function. Treatment with recombinant OSM leads to the inhibition of proliferation and changes in cellular morphology of a number of tumor cell lines derived from a wide variety of tissue types. OSM signals through cell surface receptors that contain the protein gp130. The type I receptor is composed of gp130 and LIFR, the type II receptor is composed of gp130 and OSMR.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human OSM standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human OSM standard solution and each sample be measured in duplicate.