Order Entry
United States
ContactUsLinkComponent
Human IL-17 ELISA Kit
Human IL-17 ELISA Kit
Catalog # 76236-698
CAS Number:  
Human IL-17 ELISA Kit
Catalog # 76236-698
Supplier Number:  KOA0241
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Unconjugated
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-17
  • Description:
    Human IL-17 ELISA Kit
  • Size:
    1 Kit
  • Sample Type:
    Cell culture supernates, serum and plasma(heparin, EDTA)
  • Cross Reactivity:
    There is no detectable cross-reactivity with other relevant proteins.
  • Shelf Life:
    6 months (4 °C)
  • Detection Range:
    31.2 - 2000 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76236-698
  • No. of tests:
    96 wells

Specifications

About this item

Human Interleukin-17 AccuSignal ELISA Kit.

  • Natural and recombinant human IL-17
  • There is no detectable cross-reactivity with other relevant proteins
  • Expression system for standard: E.coli; Immunogen sequence:  I20-A155

IL-17 is an inflammatory cytokine produced primarily by a unique lineage of CD4 T cells that plays critical roles in the pathogenesis of multiple autoimmune diseases. Interleukin-17 is expressed by activated T cells and is 57% identical to the 17- to 26-kD secretory glycoprotein encoded by gene 13 of the herpesvirus saimiri (HVS-13). IL17 induces nuclear factor kappa-B and the expression of IL6, intercellular adhesion molecule-1, granulocyte macrophage colony-stimulating factor,and prostaglandin E2, as well as the maturation of CD34 positive hematopoietic precursors into neutrophils. Anti-IL17 antibodies significantly inhibited osteoclast formation induced by culture media of RA synovial tissues. The standard product used in this kit is recombinant human IL-17, consisting of 136 amino acids with the molecular mass of 16KDa.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1 ml per well of the 2000 pg/ml, 1000 pg/ml, 500 pg/ml, 250 pg/ml, 125 pg/ml, 62.5 pg/ml, 31.2 pg/ml human IL-17 standard solutions into the precoated 96-well plate. Add 0.1 ml of the sample diluent buffer into the control well (Zero well). Add 0.1 ml of each properly diluted sample of human cell culture supernates, serum and plasma (heparin, EDTA) to each empty well. It is recommended that each human IL-17 standard solution and each sample be measured in duplicate.