Order Entry
United States
ContactUsLinkComponent
Mouse IL-2 ELISA Kit, Rockland Immunochemicals
Mouse IL-2 ELISA Kit, Rockland Immunochemicals
  76236-654
 :  
Mouse IL-2 ELISA Kit, Rockland Immunochemicals
  76236-654
 :  KOA0215
 :  

 

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-2
  • Description:
    Mouse IL-2 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-654

 

 

Mouse Interleukin-2 AccuSignal ELISA Kit

  • Natural and recombinant mouse IL-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A21-Q169

Interleukin-2(IL2), formerly referred to as T-cell growth factor, is a powerfully immunoregulatory lymphokine that is produced by lectin- or antigen-activated T cells. It is produced not only by mature T lymphocytes on stimulation but also constitutively by certain T-cell lymphoma cell lines. The lymphokine interleukin-2(IL-2) is responsible for autocrine cell cycle progression and regulation of immune responses. IL-2 expression in mature thymocytes and T cells has been found to be tightly controlled by monoallelic expression. IL-2 can act as a growth hormone for both B and T lymphocytes. The human gene for interleukin 2(IL2) is assigned to chromosome 4. Mouse IL-2 contains 149 amino acids in mature form with the molecular mass of 17.2KDa.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma( heparin, EDTA, citrate) to each empty well. It is recommended that each mouse IL-2 standard solution and each sample be measured in duplicate.