Order Entry
United States
ContactUsLinkComponent
Mouse IGFBP2 ELISA Kit, Rockland Immunochemicals
Mouse IGFBP2 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-628
CAS Number:  
Mouse IGFBP2 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-628
Supplier Number:  KOA0202
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IGFBP-2
  • Description:
    Mouse IGFBP2 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-628

Specifications

About this item

Mouse IGFBP2 AccuSignal ELISA Kit

  • Natural and recombinant mouse IGFBP2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E35-Q305

Insulin-like growth factor-binding protein 2(IGFBP2), also called IBP2, is a protein that in humans is encoded by the IGFBP2 gene. It is mapped to 2q35. By structural analysis, it showed that the IGFBP2 gene consists of 4 exons with 3 introns of lengths 27.0, 1.0, and 1.9 kb. IGFBP2 secreted by metastatic cells recruits endothelia by modulating IGF1 mediated activation of the IGF type-I receptor on endothelial cells. The IGFBP2/IGF1/IGF1R and GAS6/MERTK signaling pathways are regulators of cancer-mediated endothelial recruitment.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml mouse IGFBP2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IGFBP2 standard solution and each sample be measured in duplicate.