ContactUsLinkComponent
Intracellular ThiolEZ™ Cell Survival Assay Kit, Rockland Immunochemicals
Intracellular ThiolEZ™ Cell Survival Assay Kit, Rockland Immunochemicals
Catalog # 75929-182
CAS Number:  
Intracellular ThiolEZ™ Cell Survival Assay Kit, Rockland Immunochemicals
Catalog # 75929-182
Supplier Number:  KLD-004
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Intracellular ThiolEZ™ Cell Survival Assay Kit
  • Size:
    1 Kit
  • Environmentally Preferable:
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    75929-182
  • Supplier no.:
    KLD-004
Specifications
About this item
Intracellular ThiolEZ™ Cell Survival Assay Kit
  • Intracellular ThiolEZ™ Cell Survival Assay Kit contains enough reagents for 100 assays using a 6-well plate.
Intracellular ThiolEZ™ Cell Survival Assay Kit is an easy to use assay for measurement of thiols in cells; it measures total intracellular thiols including glutathione and cysteine. Glutathione is a natural antioxidant and a major thiol that limits cellular damage in healthy tissues during oxidative stress. It is also involved in repairing damage induced by cancer drugs and radiation and in the detoxification of several commonly used cancer chemotherapeutic drugs. Cysteine, which is a precursor for glutathione synthesis, is a further thiol present in cells. Cysteine is 10 times less abundant than the glutathione. Intracellular thiolEZ™ is ideal for investigators involved in aging research, oxidative stress, antioxidant screening, chemotherapy response and toxicology.
Intracellular ThiolEZ™ Cell Survival Assay Kit contains enough reagents for 100 assays using a 6-well plate.