Bovine/Calf Serum Non-Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Certificates
About this item
Non-Sterile Serum
- prepared by removal of fibrinogen through clot formation
- separated from cellular components and purified through centrifugation and filtration techniques
Bovine serum is used as a component of bioassays, immunoassays or enzyme assays. Bovine serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Bovine serum is also available as a sterile preparation for use as a cell culture supplement. Bovine Serum is ideal for investigators in Cancer, Immunology and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Specifications
- Catalog No:
- 10799-896
- 10799-898
- 10799-900
- 10799-902
- Shipping Conditions:
- Dry Ice
- Dry Ice
- Dry Ice
- Dry Ice
Specifications
Frequently Bought Together