Order Entry
United States
ContactUsLinkComponent
Sheep Serum Sterile, Rockland Immunochemicals
Sheep Serum Sterile, Rockland Immunochemicals
Catalog # RLD111-00-1000
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Sheep Serum Sterile, Rockland Immunochemicals
Catalog # RLD111-00-1000
Supplier Number:  D111-00-1000
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Sheep Serum Sterile
  • Size:
    1 L
  • Shipping Conditions:
    Dry Ice
  • Cat. No.:
    RLD111-00-1000
  • Supplier no.:
    D111-00-1000

Specifications

About this item

Sterile Serum

  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques

Sheep Serum is used as a supplement to cell culture media. Sheep Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Sheep Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Sheep Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.

pH: normal

Immunoelectrophoresis: normal

Hemoglobin: normal

IgG Concentration: normal