About this item
Sterile Serum
- 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
- prepared by removal of fibrinogen through clot formation
- separated from cellular components and purified through centrifugation and filtration techniques
Guinea Pig Serum is used as a supplement to cell culture media. Guinea Pig Serum is also suitable for use as a component of bioassays, immunoassays or enzyme assays. Guinea Pig Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation. Guinea Pig Serum is ideal for investigators in Cancer, Immunology, and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75928-670
- 75928-668
- Shipping Conditions:
- Dry Ice
- Dry Ice