ContactUsLinkComponent
Rabbit 'B' (Non-Pooled) Complement (Fresh Frozen), Rockland Immunochemicals
Rabbit 'B' (Non-Pooled) Complement (Fresh Frozen), Rockland Immunochemicals
Catalog # 75935-070
Rabbit 'B' (Non-Pooled) Complement (Fresh Frozen), Rockland Immunochemicals
Catalog # 75935-070
Supplier Number:  C308
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Rabbit ''B'' (Non-Pooled) Complement (Fresh Frozen)
  • Size:
    10 mL
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    75935-070
  • Supplier no.:
    C308
Specifications
About this item
Complement Fresh Frozen
  • prepared by removal of fibrinogen through clot formation under proper temperature conditions
  • Complement is preservative FREE
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity. Rabbit "B" Complement is suitable for CFT and SRH.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal