To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Rabbit ''C'' (5-6 Lbs.) Complement (Lyophilized) With Diluent
- Size:5 mL
- Environmentally Preferable:
- Shipping Conditions:Ambient
- Cat. No.:RLC204-0005
- Supplier no.:C204-0005
Specifications
About this item
Complement Lyophilized with DILUENT
- suitable for complement fixation tests (CFT) and Single radial haemolysis (SRH)
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal