Order Entry
United States
ContactUsLinkComponent
Anti-Strep-tag II Polyclonal Antibody
Anti-Strep-tag II Polyclonal Antibody
  75932-316
Anti-Strep-tag II Polyclonal Antibody
  75932-316
 :  600-401-H22

 

  • Antigen Symbol:
    Strep-tag II
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Size:
    100 µg
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    75932-316
  • Supplier no.:
    600-401-H22

 

 

Rabbit Anit-Strep-tag II Antibody

  • Affintiy Purified
  • ELISA, Western Blot

Strep-tag II antibody detects amino acid sequence specific for Streptavidin. Streptavidin is a tetrameric protein purified from Streptomyces avidinii. It has wide use in numerous molecular biological protocols dues to its strong affinity for biotin. The original Strep-tag is a nine amino acid peptide with high specificity and affinity towards streptavidin. The addition of this tag to the C-terminus of recombinant proteins allowed the simple purification of protein by use of affinity columns. It was re-engineered (and re-named Strep-tag II) to allow it to also placed at the N-terminus of recombinant proteins. Anti-Strep-tag II antibodies are ideal for investigators involved in GFP and Epitopes research.

Anti-Strep-tag II Antibody is suitable for use in E and WB. Specific conditions for reactivity should be optimized by the end user.