Order Entry
United States
ContactUsLinkComponent
 
Mouse Recombinant PDGF-BB (from E. coli)
 
Mouse Recombinant PDGF-BB (from E. coli)

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    Store vial at 4 °C prior to restoration. Dilute only prior to immediate use. Maintain sterility. This product DOES NOT contain preservative. DO NOT VORTEX. We recommend adding a carrier protein such as HSA or BSA to 0.1% (i.e. 1.0 mg/mL). For best results aliquot contents and freeze at –20 °C or colder. Avoid cycles of freezing and thawing. Centrifuge vial before each opening to dislodge contents from the cap and to clarify if contents are not clear after standing at room temperature.
  • Endotoxin Content:
    Measured by kinetic LAL analysis and is typically ≤ 1 EU/μg protein.
  • Biological Activity:
    The activity is measured by the dose-dependent proliferation in mouse 3T3 fibroblasts and is typically 1-2 ng/mL.
  • Gene ID:
    P31240
  • Reconstitution Instructions:
    Restore with deionized water (or equivalent); 1.0 mL
  • Protein Synonyms:
    Proto-oncogene c-Sis|Platelet-derived growth factor B chain|Platelet-derived growth factor beta polypeptide|PDGF-2
  • Protein/Peptide Name:
    PDGF-BB
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient

 

Frequently Bought Together