Mouse Recombinant CCL8
About this item
Recombinant Mouse CCL8 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Monocyte Chemotactic protein 2 (MCP-2), also known as CCL8, is implicated in allergic responses through its ability to activate mast cells, eosinophils, and basophils. MCP-2 is known to signal through several G protein-coupled receptors including, CCR1, CCR2B, and CCR5. Recombinant mouse MCP-2 is a non-glycosylated protein, containing 74 amino acids, with a molecular weight of 8.5 kDa.
Monocyte Chemotactic Protein-2 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-2 in immunological assays.
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recombinant Mouse CCL8 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Monocyte Chemotactic protein 2 (MCP-2), also known as CCL8, is implicated in allergic responses through its ability to activate mast cells, eosinophils, and basophils. MCP-2 is known to signal through several G protein-coupled receptors including, CCR1, CCR2B, and CCR5. Recombinant mouse MCP-2 is a non-glycosylated protein, containing 74 amino acids, with a molecular weight of 8.5 kDa.
Monocyte Chemotactic Protein-2 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-2 in immunological assays.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 75933-898
- 75933-896
- 75930-044
- Protein/Peptide Name:
- CCL8
- CCL8
- CCL8
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Mouse
- Shipping Temperature:
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: 010-001-V75-0005Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.75933-898Protein/Peptide NameCCL8Protein/Peptide TypeRecombinantSpeciesMouseShipping TemperatureAmbient - Supplier Number: 010-001-V75-0020Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.75933-896Protein/Peptide NameCCL8Protein/Peptide TypeRecombinantSpeciesMouseShipping TemperatureAmbient - Supplier Number: 010-001-V75-0100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.75930-044Protein/Peptide NameCCL8Protein/Peptide TypeRecombinantSpeciesMouseShipping TemperatureAmbient
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



