About this item
Recombinant Human TARC /CCL17 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Thymus and activation regulated chemokine (TARC), also known as CCL17, is a chemokine produced by thymus tissue constitutively and activated PBMCs (mainly DCs). TARC signals through the CCR4 receptor to induce chemotaxis of Th2 cells. TARC is thought to be important in asthma and allergic diseases, along with bacterial and viral infections. Recombinant human TARC is a non-glycosylated protein, containing 71 amino acids, with a molecular weight of 8.1 kDa.
TARC Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-TARC in immunological assays.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:TARC /CCL17
- Shipping Temperature:Ambient