Recombinant Human Neurotrophin-4 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Neurotrophin-4 (NT-4) is an important member of the nerve growth factor (NGF) family of proteins along with NT-3, BDNF and NGF. Neurotrophins act in a paracrine and/or autocine manner to control neuronal numbers and dendrite outgrowth. These functions are thought to be communicated by TrkB, a receptor tryrosine kinase that NT-4 shares with BDNF. Recombinant human NT-4 is a non-glycosylated, non-covalently linked homodimer, containing two 131 amino acid chains, with a total molecular weight of 28.1 kDa.
Neurotrophin-4 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Neurotrophin-4 in immunological assays.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Source:
- Species:
- Environmentally Preferable: