Human Recombinant NGF beta
Supplier: Rockland Immunochemical
Certificates
About this item
Recombinant Human NGF beta control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Nerve Growth Factor (NGF-β) is a neurotrophic factor related to BDNF, NT-3 and NT-4. NGF-β acts through its receptor β-NGFR, and is involved in the development and maintenance of the sensory and sympathetic nervous systems. NGF-β is also involved in the growth, differentiation and survival of B lymphocytes. Human, mouse and rat proteins show cross-reactivity. Recombinant human NGF-β is a non-covalently linked homodimer, containing two 121 amino acid chains, with a total molecular weight of 27.2 kDa.
Nerve Growth Factor beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Nerve Growth Factor beta in immunological assays.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:NGF beta
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together