Human Recombinant CCL7
Supplier: Rockland Immunochemical
Certificates
About this item
Recombinant Human CCL7 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Monocyte Chemotactic Protein 3 (MCP-3), also called CCL7, is produced by macrophages and some tumor cell lines. MCP-3 signals through three different G protein-coupled receptors, CCR1, CCR2, and CCR3. CCL7 chemoattracts monocytes and can regulate macrophage function. Recombinant human MCP-3 is a non-glycosylated protein, containing 76 amino acids, with a molecular weight of 9 kDa.
Monocyte Chemotactic Protein-3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-3 in immunological assays.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:CCL7
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together