About this item
Recombinant Human IGF-II control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Insulin-like Growth Factor II, IGF-II, is a major growth hormone made by Theca cells during gestation. While IGF-II is known to engage the IGF-I receptor (IGF1R) to mediate embryonic growth, IGF-II is also known to bind the IGF-II receptor (IGF2R). IGF2R is thought to be signaling dead receptor that acts as a sink by binding up free IGF-II. Recombinant human IGF-II is non-glycosylated protein, containing 67 amino acids, with a molecular weight of 7.5 kDa.
Insulin-like Growth Factor II Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Insulin-like Growth Factor II in immunological assays.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:IGF-II
- Shipping Temperature:Ambient