Recombinant Human CTGF control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Connective Tissue Growth Factor (CTGF) is a member of cysteine rich regulatory proteins that are both mitogenic and chemotactic. Each protein has an Insulin-like Growth Factor (IGF)-binding domain, a thrombospondin type 1 domain and cysteine knot region. CTGF has multiple effects on development and differentiation. Recombinant human CTGF is a non-glycosylated protein, containing 98 amino acids, with a molecular weight of 11.2 kDa.
Connective Tissue Growth Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Connective Tissue Growth Factor in immunological assays.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:CTGF
- Shipping Temperature:Ambient