Orders LinkContactUsLinkComponent
HSPB1
HSPB1
HSPB1
Certificates
1 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
HSP27 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HSP27 is a member of heat shock proteins (HSPs) which are synthesized in cells in response to heat shock and other metabolic stresses and provide a transient state of thermotolerance. HSP27 plays a major role in the increased thermal resistance acquired by cells after exposure to HSP inducers (1). The level of HSP27 phosphorylation is significantly elevated after exposure of cells to heat shock, sodium arsenite, IL-1 and TNF-a. MAPKAPK2 and MAPKAPK3 are both activated by these conditions and can phosphorylate HSP27 on serine residues (2). HSP27 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Cancer, Cellular Stress, and p38 Pathway research.
HSP27 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. HSP27 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~52kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 75928-740
  • 75928-742
  • Protein/Peptide Name:
  • HSPB1
  • HSPB1
  • Shipping Temperature:
  • Dry Ice
  • Dry Ice
Specifications
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 20 µg
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $394.55
    $394.55
    Each (20µG)
       $394.55
    Minimum order quantity is 1
    Supplier Number: 009-001-234S
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    75928-740
    Protein/Peptide Name
    HSPB1
    Shipping Temperature
    Dry Ice
  • 50 µg
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    75928-742
    Protein/Peptide Name
    HSPB1
    Shipping Temperature
    Dry Ice