Order Entry
United States
ContactUsLinkComponent
LL-37, Fluorescein Conjugated, FITC (Fluorescein Isothiocyanate)
LL-37, Fluorescein Conjugated, FITC (Fluorescein Isothiocyanate)
Catalog # RL000-002-M35
CAS Number:  
LL-37, Fluorescein Conjugated, FITC (Fluorescein Isothiocyanate)
Catalog # RL000-002-M35
Supplier Number:  000-002-M35
CAS Number:  

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Size:
    1 mg
  • Protein/Peptide Name:
    LL-37, Fluorescein conjugated
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    RL000-002-M35
  • Supplier no.:
    000-002-M35

Specifications

About this item

Cathelicidin (LL-37) Control Peptide

  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot

LL-37 is a cleaved antimicrobial residue peptide of hCAP18 (human cationic antimicrobial protein, 18kDa) in the family of cathelicidin. Cathelicidins are predominatly found in the peroxidase negative granules of neutophils. And it has been shows broad antimicrobial activity towards gram negative and gram positive bacteria.

LL-37, N-5FAM Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.