Order Entry
United States
ContactUsLinkComponent
MAD2L2 Peptide
MAD2L2 Peptide
Catalog # RL000-001-470
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
MAD2L2 Peptide
Catalog # RL000-001-470
Supplier Number:  000-001-470
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Size:
    50 µg
  • Environmentally Preferable:
  • Protein/Peptide Name:
    MAD2L2 Peptide
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    RL000-001-470
  • Supplier no.:
    000-001-470

Specifications

About this item

MAD2L2 Control Peptide

  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot

MAD2L2 is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. MAD2L2 is a homolog of MAD2L1.

Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay.