ContactUsLinkComponent
QC Sets and Panels: Helix Elite, Microbiologics
QC Sets and Panels: Helix Elite, Microbiologics
Catalog # 76299-456
CAS Number:  
undefined
QC Sets and Panels: Helix Elite, Microbiologics
Catalog # 76299-456
Supplier Number:  8204
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    BD Max Enteric Parasite Control Panel
  • Cat. No.:
    76299-456
Specifications
About this item
All the required QC strains for your lab’s microbial detection/identification system, already combined in one convenient panel.
  • One simple catalog number for fast, easy ordering
  • Protects your reputation by providing documentation and assurance that the laboratory can consistently produce accurate results
  • Ensures testing procedures, equipment and materials are working properly
  • Ready-to-use format saves time and money
  • Online Certificate of Analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
  • Technical Support experts available for guidance
Each instrument-specific QC set or panel contains the manufacturer recommended quality control strains for a particular instrument or test in an easy-to-use format.
Each regulation-specific QC set or panel contains the regulation-specific (e.g. CLSI® or EUCAST) recommended quality control strains for a particular test in an easy-to-use format.
All the required QC strains for your lab’s microbial detection/identification system, already combined in one convenient panel. We offer a wide range of bundled QC Sets and Panels for the most commonly used technologies and standards. Each set or panel contains the manufacturer-recommended quality control strains for a particular instrument or test method in a simple, ready-to-use format.