Order Entry
United States
ContactUsLinkComponent
Premiere 2-in-1 Low Speed Centrifuge
Premiere 2-in-1 Low Speed Centrifuge
Catalog # 470303-826
Supplier:  C & A Scientific
undefined
Premiere 2-in-1 Low Speed Centrifuge
Catalog # 470303-826
Supplier:  C & A Scientific
Supplier Number:  XC-2500

Specifications

  • Description:
    2-in-1 Low Speed Centrifuge
  • Max. RCF:
    1828 ×g
  • Speed:
    1000 - 4000 rpm
  • Cat. No.:
    470303-826

Specifications

About this item

Programmable settings with bright and easy-to-read LCD display.

  • One touch panel for quick and easy timer settings
  • Fixed Angled Rotor with adapters for 6×15 ml or 50 ml tube holders
  • Built-in safety feature keeps lid closed while rotor is in use
  • Brushless motor for long lasting operation

With the versatile power of 2 centrifuges in one, the XC-2500 can spin 15 and 50 ml tubes — saving your customers diagnostic time, equipment cost, and counter-top space!

Used in clinical settings for urine spinning, whole cell separation, cell washing, whole blood separation, pharmaceuticals, Platelet Rich Plasma (PRP), and Platelet Rich Fibrin (PRF).