OIML Class E2 Individual Standard Line Weights, METTLER TOLEDO®
Supplier: Mettler Toledo
Certificates
About this item
Weights made from austenitic stainless steel are protected in robust and easy to clean plastic boxes.
- Low magnetic susceptibility
- Monobloc
- Density: 8 kg/dm³
- High corrosion resistance
- Easy to clean
The one-piece construction and high-gloss polished surface offer best long term stability.
Certifications: Calibration at ISO 17025:2005 A2LA accredited lab Worthington, OH.
Frequently Bought Together