Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Surface Protectors
Bench Protectors
Mat Sheeting, Purestat Technology
Mat Sheeting, Purestat Technology
Mat Sheeting, Purestat Technology
Catalog # 76245-332
Supplier:
Purestat Technology
Expand image
Mat Sheeting, Purestat Technology
Catalog # 76245-332
Supplier:
Purestat Technology
Supplier Number:
48 X 500 X .00
Specifications
Color:
Blue
Description:
Mat sheeting
Thickness:
4 mil.
Cat. No.:
76245-332
Specifications
About this item
Product Details
Product Details