Metal Expansion Wristband, SCS, Desco Industries
Catalog # 76239-632
Supplier: Desco
Specifications
- Description:Metal wristband, small
- Cat. No.:76239-632
Specifications
About this item
Metal Expansion Wristband made from stainless steel, the durable premium performance metal wristband creates a reliable electrical contact to the user’s wrist.
- Durable and comfortable
Each link is covered with an insulative plastic cap. Use with single-wire coil cord with 4 mm female socket snap.
Certifications: Meets ANSI/ESD S20.20 tested per ANSI/ESD S1.1 and ESD TR53.