ContactUsLinkComponent
Anticon® Gold StandardWeight™ Series Cleanroom Wipers, Contec®
Anticon® Gold StandardWeight™ Series Cleanroom Wipers, Contec®
Supplier:  Contec
Certificates
undefined
Anticon® Gold StandardWeight™ Series Cleanroom Wipers, Contec®
Supplier:  Contec
Specifications
  • Catalog No:
  • 76239-616
  • 76239-552
  • Basis Weight:
  • 120 g/m²
  • 120 g/m²
  • Absorbency:
  • 417 ml/m²
  • 417 ml/m²
  • Absorbency - Rate:
  • 0.2 sec.
  • 0.2 sec.
  • Extractables:
  • 0.0029 g/m² using deionized water, 0.0227 g/m² using isopropyl alcohol
  • 0.0029 g/m² using deionized water, 0.0227 g/m² using isopropyl alcohol
  • Fibers (>100 µm):
  • 0.0037/cm²
  • 0.0037/cm²