Soldering Handle with Reinforced Cable, JBC
Catalog # 76252-262
Supplier: JBC TOOLS USA INC. TE
Specifications
- Description:General Purpose Soldering Handle with reinforced cable
- Length:1.5 m
- Environmentally Preferable:
- Cat. No.:76252-262
Specifications
About this item
This soldering iron handle for general purposes features a soft foam grip which provides thermal insulation. It is also perfect for SMD components.
- Ergonomic handle
The short distance tip-to-grip ensure greater results and comfort.
Delivery: Supplied with reinforced cable for jobs with high power requirements.